missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PURB Polyclonal Antibody
GREENER_CHOICE

Product Code. 17158173
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17158173 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17158173 Supplier Invitrogen™ Supplier No. PA5110245

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GGLYGELPEG TSITVDSKRF FFDVGCNKYG VFLRVSEVKP SYRNAITVPF KAWGKFGGAF CRYADEMKEI QERQRDKLYE RRGGGSGGGE ESEGEEVDED.

This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PURB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PURB
Gene Accession No. Q96QR8
Gene Alias 2310015K15Rik; AA114818; Cager-2; D11Bwg0414e; LOW QUALITY PROTEIN: transcriptional activator protein Pur-beta; purb; PurB beta; purb protein; purbb; PURBETA; pur-beta; purine rich element binding protein B; purine-rich element binding protein B; purine-rich element binding protein Bb; purine-rich element-binding protein B; purine-rich single-stranded DNA-binding protein beta; transcription factor Pur-beta; transcriptional activator protein Pur-beta; Vascular actin single-stranded DNA-binding factor 2 p44 component; zgc:65916
Gene Symbols PURB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 213-312 of human PURB (NP_1500931).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5814
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.