Learn More
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 10 to 15 in. wide
GREENER_CHOICE
Product Code. 30042304 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Size:
10 x 15
10 x 18
10 x 20
10 x 22
10 x 24
10 x 30
10 x 36
10 x 48
10 x 56
10 x 62
12 x 12
12 x 14
12 x 16
12 x 18
12 x 20
12 x 22
12 x 24
12 x 28
12 x 30
12 x 32
12 x 34
12 x 36
12 x 40
12 x 42
12 x 46
12 x 48
14 x 14
14 x 16
14 x 18
14 x 20
14 x 24
14 x 26
14 x 36
14 x 38
14 x 40
14 x 48
15 x 26
15 x 43
Unit Size:
Pack of 250
38 product options available for selection
Product selection table with 38 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30042304 14 x 24 Pack of 250
30042225 14 x 20 Pack of 250
30042317 14 x 18 Pack of 250
30042243 14 x 16 Pack of 250
30042278 14 x 14 Pack of 250
30042299 12 x 48 Pack of 250
30042280 12 x 46 Pack of 250
30042203 12 x 42 Pack of 250
30042179 12 x 40 Pack of 250
30042244 12 x 36 Pack of 250
30042321 12 x 34 Pack of 250
30042207 12 x 32 Pack of 250
30042205 12 x 30 Pack of 250
30042277 12 x 28 Pack of 250
30042315 12 x 24 Pack of 250
30042231 12 x 22 Pack of 250
30042188 12 x 20 Pack of 250
30042189 12 x 18 Pack of 250
30042257 12 x 16 Pack of 250
30042255 12 x 14 Pack of 250
30042216 12 x 12 Pack of 250
30042248 10 x 62 Pack of 250
30042191 10 x 56 Pack of 250
30042251 10 x 48 Pack of 250
30042270 10 x 36 Pack of 250
30042220 10 x 30 Pack of 250
30042228 10 x 24 Pack of 250
30042212 10 x 22 Pack of 250
30042246 10 x 20 Pack of 250
30042184 10 x 18 Pack of 250
30042206 10 x 15 Pack of 250
30042201 15 x 43 Pack of 250
30042263 15 x 26 Pack of 250
30042208 14 x 48 Pack of 250
30042302 14 x 40 Pack of 250
30042281 14 x 38 Pack of 250
30042215 14 x 36 Pack of 250
30042193 14 x 26 Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 38 options available.
38 options
Product Code. 30042304 Supplier Keystone Cleanroom Products Supplier No. 6STKRP1424SS
Only null left
Add to basket
Add to basket
PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.
  • Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
  • Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
  • Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
  • Fast and consistent for equipment preparation area
  • Resists tearing and punctures
  • Superior microbial barrier
  • Manufactured and packaged in a certified ISO 6 cleanroom
  • Made in the USA
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type Safe Seal
Color Clear, White
Disposable Single-use
For Use With (Application) Steam Sterilization of Pharmaceutical Equipment
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Size 14 x 24
Temperature Range (Metric) 121 to 125 C
Thickness (English) 5.3 mil
Length (English) 24 in.
Width (English) 14 in.
Product Type Autoclave Bag
Length (Metric) 61 cm
Width (Metric) 35.6 cm
Thickness (Metric) 5.3 mil