Learn More
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 21 to 30 in. wide
GREENER_CHOICE
Product Code. p-200049745 Shop All Keystone Cleanroom Products Products
Click to view available options
Size:
21 x 34
22 x 24
22 x 26
22 x 40
22 x 60
24 x 24
24 x 30
24 x 36
24 x 40
24 x 48
24 x 52
24 x 62
26 x 28
26 x 34
26 x 46
28 x 28
28 x 32
28 x 34
28 x 40
28 x 44
30 x 16
30 x 20
30 x 28
30 x 30
30 x 36
30 x 60
Unit Size:
Pack of 100
Pack of 250
Product Code. 30042300
Brand: Keystone Cleanroom Products 6STKRP3060SS
Only null left
Add to basket
Add to basket
PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.
  • Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
  • Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
  • Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
  • Fast and consistent for equipment preparation area
  • Resists tearing and punctures
  • Superior microbial barrier
  • Manufactured and packaged in a certified ISO 6 cleanroom
  • Made in the USA
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type Safe Seal
Color Clear, White
Disposable Single-use
For Use With (Application) Steam Sterilization of Pharmaceutical Equipment
Material Tyvek
Sterility Non-sterile
Thickness (Metric) 5.3 mil
Latex Free Yes
Size 30 x 60
Temperature Range (Metric) 121 to 125 C
Thickness (English) 5.3 mil
Length (English) 60 in.
Length (Metric) 152.4 cm
Width (English) 30 in.
Width (Metric) 76.2 cm
Product Type Autoclave Bag