Learn More
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 32 to 62 in. wide
GREENER_CHOICE
Product Code. 30042221 Shop All Keystone Cleanroom Products Products
Click to view available options
Size:
32 x 32
32 x 38
32 x 48
34 x 12
34 x 34
34 x 56
36 x 24
36 x 28
36 x 36
36 x 42
36 x 46
36 x 48
36 x 60
36 x 64
36 x 72
38 x 38
40 x 24
40 x 40
40 x 48
40 x 54
40 x 60
40 x 72
42 x 42
42 x 44
44 x 16
44 x 50
48 x 48
48 x 64
50 x 54
62 x 50
Unit Size:
Pack of 100
Pack of 125
Pack of 250
Product Code. 30042221
Brand: Keystone Cleanroom Products 6STKRP4864SS
Only null left
Add to basket
Add to basket
PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.
  • Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
  • Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
  • Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
  • Fast and consistent for equipment preparation area
  • Resists tearing and punctures
  • Superior microbial barrier
  • Manufactured and packaged in a certified ISO 6 cleanroom
  • Made in the USA
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type Safe Seal
Color Clear, White
Disposable Single-use
For Use With (Application) Steam Sterilization of Pharmaceutical Equipment
Material Tyvek
Sterility Non-sterile
Thickness (Metric) 5.3 mil
Latex Free Yes
Size 48 x 64
Temperature Range (Metric) 121 to 125 C
Thickness (English) 5.3 mil
Length (English) 64 in.
Length (Metric) 162.6 cm
Width (English) 48 in.
Width (Metric) 121.9 cm
Product Type Autoclave Bag