Description
- Flat style caps for 8-well strips
- ATP, RNase/DNase and Endotoxin-free (Non-pyrogenic)
- Polypropylene USP Class VI resin
- Made in USA
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2015, FDA CFR21 GMP |
| Color | Clear |
| Volume (Metric) | 0.2 mL |
| Quantity | 125 Caps/Bag, 10 Bags/Cs. |
| Product Line | PurePlus |
| Sterility | Non-sterile |
| Material | Polypropylene |
| RNase Free | Yes |
| Closure Type | Flat Cap |
| No. of Wells | 8 Wells |
| Show More |