Learn More
Labcon™ PurePlus™ Solid Frame PCR Plates
GREENER_CHOICE
Product Code. p-200112174
Click to view available options
Array:
12 x 8
16 x 24
24 x 16
No. of Wells:
96
384
Volume (Metric) Well:
25 μL
0.1 mL
0.2 mL
Unit Size:
Pack of 100
Product Code. 30280319
Brand: Labcon™ 39855200009
Only null left
Add to basket
Add to basket
Labcon Solid Frame PCR plates are made with two materials to improve the fit in thermocyclers and allow for rapid cycling.
  • Made with a rigid polycarbonate frame enclosing polypropylene reaction wells
  • Available in styles to fit major cyclers and are freezable (-80°C)
  • Choose from 96 or 384 well capacity
Specifications
Barcode No Barcode
Certifications/Compliance ISO 9001:2015, FDA CFR 21 GMP
Format SBS Standard
Array 12 x 8
Autoclavable Not Autoclavable
Packaging 10 Plates/Pk., 10 Pk./Cs.
Color Clear
Disposable Single-use
Product Type PCR plate
Skirt Style Full skirt
Material Polycarbonate
Volume (Metric) Well 0.1 mL
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) PCR
High-throughput Compatibility Yes
RNase Free Yes
Well Color Clear
Fit for Purpose PCR Ready