missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purine Nucleoside Phosphorylase/PNP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99935-50ul
This item is not returnable.
View return policy
Description
Purine Nucleoside Phosphorylase/PNP Polyclonal antibody specifically detects Purine Nucleoside Phosphorylase/PNP in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| Purine Nucleoside Phosphorylase/PNP | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate | |
| EC 2.4.2.1, FLJ94043, FLJ97288, Inosine phosphorylase, MGC117396, MGC125915, MGC125916, NPFLJ97312, nucleoside phosphorylase, PRO1837, PUNP, purine nucleoside phosphorylase, purine-nucleoside:orthophosphate ribosyltransferase | |
| Produced in rabbits immunized with purified, recombinant Human Purine Nucleoside Phosphorylase/PNP (Accession#: P00491; Met1-Ser289) | |
| 50 μg | |
| Neuroscience | |
| 4860 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction