missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PURL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97687-50ul
This item is not returnable.
View return policy
Description
PURL Polyclonal antibody specifically detects PURL in Human samples. It is validated for Western Blot
Specifications
| PURL | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FGAM synthase, FGAMS, FGAR amidotransferase, formylglycinamide ribotide synthetase, KIAA0361, phosphoribosylformylglycinamidine synthase, PURL | |
| Produced in rabbits immunized with E. coli-derived Human PURL fragment. | |
| 50 μg | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 5198 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction