Learn More
LSECtin/CLEC4G Antibody (845404), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2947B
Product Code. 18678343
  • € 670.00 / 0.10mL
Description
LSECtin/CLEC4G Monoclonal antibody specifically detects LSECtin/CLEC4G in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LSECtin/CLEC4G
Monoclonal
Biotin
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
339390
Store at 4°C in the dark.
IgG2a
Flow Cytometry
845404
Flow Cytometry
C-type lectin domain family 4 member G, C-type lectin domain family 4, member G, C-type lectin superfamily 4, member G, DTTR431, liver and lymph node sinusoidal endothelial cell C-type lectin, LP2698, LSECtin, UNQ431
NS0 mouse myeloma cell line transfected with LSECtin/CLEC4G, Ser54-Cys293, Accession # Q6UXB4
0.1 mL