Learn More
Novus Biologicals™ Purple Hematoxylin Solution
Shop All Bio Techne Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| For Use With (Application) | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Quantity | 100 mL |
| Technique | Preparation of Working Solutions Purple Hematoxylin is ready to use and does not require any preparation. Protocol Recommendations Counterstain 1. Rinse slides with deionized or distilled water. 2. Place slides into Purple Hematoxylin for 30-60 seconds. 3. Rinse slides in deionized or distilled water until all free hematoxylin has been removed. 4. Bluing the slides in an alkaline solution is not required and is not recommended. |