Description
- Spray head for Pursept™ A Xpress 1L bottle provides effective distribution.
- Adjustablle nozzle determines amount of liquid dispensed.
Specifications
Specifications
| Product Type | Spray Head |
| Color | White |
| Description | Fits 1L bottle |
| For Use With (Equipment) | Pursept A Xpress Surface Disinfecting Spray |
| Quantity | 1 Ea. |
Safety and Handling
Product Identifier
- pursept A Xpress
- MIXTURE LIST-Contain: Ethanol