missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 391.65 - € 590.10
Specifications
| Antigen | PUS1 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18225012
|
Novus Biologicals
NBP2-56899 |
100 μL |
€ 624.00 € 590.10 / 100µL Save € 33.90 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674936
|
Novus Biologicals
NBP2-56899-25ul |
25 μL |
€ 415.00 € 391.65 / 25µL Save € 23.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PUS1 Polyclonal specifically detects PUS1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| PUS1 | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 80324 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LKVWLIDDILEKINSHLPSHIRILGLKRVTGGFNSKNRCDARTYCYLLPTFAFAHKDRDVQDETYRLSAETLQQVNRLLACYKGTHNFHNFTSQ | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 5.4.99.12, MGC11268, mitochondrial tRNA pseudouridine synthase A, MLASA1, pseudouridylate synthase 1, tRNA pseudouridine synthase A, mitochondrial, tRNA pseudouridylate synthase I, tRNA uridine isomerase I, tRNA-uridine isomerase I | |
| PUS1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title