missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PUS1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA562448

Product Code. 15750242

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PQDPSACRYI LEMYCEEPFV REGLEFAVIR VKGQSFMMHQ IRKMVGLVVA IVKGYAPESV LERSWGTEKV DVPKAPGLGL VLERVHFEKY NQRFG Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

The PRKDC gene encodes the catalytic subunit of a nuclear DNA-dependent serine/threonine protein kinase (DNA-PK). The second component is the autoimmune antigen Ku, which is encoded by the G22P1 gene on chromosome 22q. On its own, the catalytic subunit of DNA-PK is inactive and relies on the G22P1 component to direct it to the DNA and trigger its kinase activity; PRKDC must be bound to DNA to express its catalytic properties.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PUS1
Polyclonal
Unconjugated
PUS1
A730013B20Rik; mitochondrial tRNA pseudouridine synthase A; MLASA1; MNCb-0873; MPUS1; mPus1p; PP8985; pseudouridine synthase 1; pseudouridylate synthase 1; Pus1; pus1 {ECO:0000312; RGD:1311871}; tRNA pseudouridine synthase A; tRNA pseudouridine synthase A, mitochondrial; tRNA pseudouridine(38-40) synthase; tRNA pseudouridylate synthase I; tRNA uridine isomerase I; tRNA-uridine isomerase I
Rabbit
Antigen affinity chromatography
RUO
80324
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9Y606
PUS1
Recombinant protein corresponding to Human PUS1. Recombinant protein control fragment (Product #RP-97865).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.