missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUS7 Antibody (OTI5F12), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73733PECY55
This item is not returnable.
View return policy
Description
PUS7 Monoclonal specifically detects PUS7 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications
| PUS7 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| PUS7 | |
| Full length recombinant protein of human PUS7 (NP_061915) produced in HEK293T cells. | |
| 0.1 mL | |
| 54517.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI5F12 | |
| Flow Cytometry | |
| EC 5.4.99.-, FLJ20485, KIAA1897, MGC17720, pseudouridylate synthase 7 homolog, pseudouridylate synthase 7 homolog (S. cerevisiae) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction