missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Puumala Virus Glycoprotein Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41260AF405
This item is not returnable.
View return policy
Description
Puumala Virus Glycoprotein Polyclonal antibody specifically detects Puumala Virus Glycoprotein in Virus samples. It is validated for ELISA
Specifications
| Puumala Virus Glycoprotein | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| ELISA | |
| Alexa Fluor 405 | |
| Envelopment polyprotein, Gc, Glycoprotein C, Glycoprotein G1, Glycoprotein G2, Glycoprotein N, Gn, M polyprotein, Precursor, PUUVsMgp1 | |
| Antibody was raised against a 19 amino acid peptide near the amino terminus of the Puumala virus glycoprotein. The immunogen is located within amino acids 90 - 140 of Puumala Virus Glycoprotein. Amino Acid Squence: TQASSTSFQTKSSEVNLRG | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 2943082 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction