Learn More
Buerkle™ PVC Conical Cap for OTAL™ Pumps
Spare part for OTAL™ Pump. Helps to provide airtight seal.
Brand: Buerkle™ 5000-0013
22.09 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
- Spare part for Buerkle™ OTAL™ Pump
- Helps to provide an airtight seal
Specifications
Replacement Part | |
Conical bung | |
PVC | |
No |
Red | |
Each | |
OTAL Pump with Tube Diameter 18 mm | |
Conical |