Learn More
Buerkle™ PVC Conical Cap for OTAL™ Pumps
Spare part for OTAL™ Pump. Helps to provide airtight seal.
€ 24.10
Specifications
Description | Replacement Part |
---|---|
Autoclavable | No |
Color | Red |
Material | PVC |
Product Type | Conical bung |
Description
- Spare part for Buerkle™ OTAL™ Pump
- Helps to provide an airtight seal
Specifications
Replacement Part | |
Red | |
Conical bung | |
Each |
No | |
PVC | |
Conical |