Learn More
Gilson™ PVC Tubing for MF1 and MF4 pump head
Product Code. p-8001660 Shop All Gilson Products
Click to view available options
Diameter (Metric) Inner:
0.25 mm
0.38 mm
0.51 mm
1.02 mm
1.52 mm
2.06 mm
2.79 mm
3.18 mm
Flow Rate:
0.574mL/min.
1.22mL/min.
1.9mL/min.
14mL/min.
25mL/min.
39mL/min.
47mL/min.
6mL/min.
Unit Size:
3m
Product Code. 10464712
Brand: Gilson™ F117952
Only null left
Add to basket
Add to basket
For use with MF1 and MF4 pump head
  • PVC
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 0.25 mm
Material PVC
Diameter (Metric) Outer 2.1 mm
Overall Length (Metric) 300 cm
For Use With For MF1 and MF4 pump heads
For Use With (Application) general use
Packaging Quantity 1 Each
Flow Rate 0.574mL/min.
Gas Permeability Low
Color Transparent
Max. Operating Temperature (Metric) 94°C
Product Type Extension Pump Tubing