Learn More
Cole-Parmer PVC Fuel and Lubricant Tubing
Product Code. 30053300 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1/2 in.
1/4 in.
1/8 in.
2/25 in.
3/16 in.
3/8 in.
5/16 in.
7/16 in.
Diameter (English) Outer:
1/2 in.
1/4 in.
3/4 in.
3/8 in.
5/16 in.
7/16 in.
7/50 in.
9/16 in.
Unit Size:
7.6m
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Diameter (English) Outer unitSize
30053300 1/8 in. 1/4 in. 7.6m
30053384 3/16 in. 5/16 in. 7.6m
30054304 2/25 in. 7/50 in. 7.6m
30054361 3/8 in. 1/2 in. 7.6m
30053137 5/16 in. 7/16 in. 7.6m
30053794 1/4 in. 3/8 in. 7.6m
30053950 1/2 in. 3/4 in. 7.6m
30053459 7/16 in. 9/16 in. 7.6m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 30053300 Supplier Cole-Parmer Supplier No. 5012135
Only null left
Add to basket
Add to basket
Specifically formulated to resist hardening and swelling
Resist hydrocarbons, avoid swelling, and reduce cracking. Safely transfer chemicals and petrochemicals including gasoline, kerosene, oils and cutting compounds. Protects against leaching and product contamination. Ideal for applications ranging from small engine fuel lines to coolant transfer. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Specifications
Diameter (English) Inner 1/8 in.
Material PVC
Diameter (English) Outer 1/4 in.
Overall Length (English) 25 ft.
Color Yellow