Learn More
Welch Ilmvac™ PVC Hose with Support Spiral
Shop All Welch Ilmvac Products
Click to view available options
Diameter (Metric) Inner:
16 mm
Unit Size:
1m
Specifications
Specifications
| Diameter (Metric) Inner | 16 mm |
| Material | PVC |
| Diameter (Metric) Outer | 23 mm |
| Overall Length (Metric) | 100 cm |
| Packaging Quantity | 1 Each |
| Product Type | Hose with Support Spiral |