Learn More
Brady™ PVC Safety Signs
Product Code. 15143289 Shop All Brady Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15143289 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15143289 Supplier Brady™ Supplier No. 814289
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
Certifications/Compliance ISO 7010
Material PVC
Quantity 1 Ea.
Description Polyvinyl Chloride
For Use With (Application) Indoor & Outdoor
Height (Metric) 203 mm
Max. Temperature (Metric) 70°C
Thickness 0.369 mm
Width (Metric) 203 mm
Temperature Range (Metric) -20 °C-70 °C
Shape Square
Languages Global
Displayed Text Emergency exit (left)
Product Type Safety Sign
Color White on Green
Show More Show Less
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.