Learn More
Buerkle™ PVC Standard Bung
Shop All Bürkle Products
Click to view available options
Height (Metric):
30 mm
Unit Size:
Each
Description
- PVC, soft, red
Specifications
Specifications
| Height (Metric) | 30 mm |
| Length (Metric) | 28 mm |
| Width (Metric) | 34 mm |
| Color | Red |