Learn More
Masterflex™ PVDF Barbed NPT Male Pipe Adapter Elbows
Designed for quick and easy assembly without tools
Brand:  Masterflex™ 30704-34
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11876723
  • € 52.96 / Pack of 10
Estimated Shipment: 16-08-2021
Add to basket
Description
Description
  • Ideal for low-pressure applications
  • Chemical resistant
  • Available in a variety of sizes
  • Economical barbed fittings easily connect fluid paths without the use of tools
  • Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Specifications
Translucent
Autoclavable
1 3/8 in.
3/8 in.
3/8 in.
Autoclave or Ethylene Oxide
1 9/32 in.
1/8 NPT(M) x 3/8 in. ID
PVDF
Barbed NPT Male Pipe Adapter Elbows
3/8 in.
1/8 in.
1/8 in.