Learn More
Saint-Gobain Performance Plastics PVDF Clamp Flow Meter
Beskrivelse
- Infrared (IR) turbine rotor reflection results in accurate and repeatable flow measurements
- USP Class VI PVDF material can be gamma sterilized up to 50kGy
- Each sensor is pre-calibrated to reach <1% accuracy
- High resolution square wave output
- Performs an easy and fast exchange of the wetted parts
- Small housing with screw gap to mount onto your device or system
- Tubes can be sterilized up to 150°C (302°F)
- Reduces down-time and prevents cross-contamination
Tekniske data
Tekniske data
| For Use With (Equipment) | Flowmeter |
| Type | Flow Sensor (Clamp-mount) |
| Includes | 4.5 PVDF, 1/8 NPT Clamp |
| Airflow Rate | 0.03 to 2 L/min. |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?