Learn More
Cole-Parmer™ PVDF Manifold, 4 port
Expand your flow and line capacity with these manifolds. Choose the number of ports and connections to suit your needs.
Brand: Cole-Parmer™ 31523-39
Additional Details : Weight : 0.80000kg
Description
- Constructed of durable PVDF.
- Ideal for applications requiring excellent chemical compatibly.
- Available in a variety of configurations
Specifications
Manifold | |
4 1/8 in. | |
-65°F | |
3/8 in. x 1/4 in. | |
1/4 in. NPT(F) | |
1.25 in. | |
1.25 in. | |
230°F |
PVDF | |
10.48 cm | |
-54°C | |
3/8 in. NPT(F) | |
3.18 cm | |
3.18 cm | |
110 psi | |
110°C |