Learn More
Cole-Parmer™ PVDF Needle Valves
Click to view available options
:
Connections: 0.5in.
Specifications
Specifications
| Material | PVDF |
| For Use With | 0.5 in. Tubing |
| Min. Operating Temperature (Metric) | 150°C |
| Packaging Quantity | 1 Pack |