Learn More
Masterflex™ PVDF NPT Male Adapter to Hose Barb
Connect fluid paths without the use of tools
Brand:  Masterflex™ 30704-17
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11806703
  • € 56.51 / Pack of 10
Estimated Shipment
26-05-2021
Add to basket
Description
Description
  • Quick and easy assembly without tools
  • Just slip your flexible tubing or hose onto the barb to create a secure seal
  • Chemical resistant
  • Available in a variety of sizes
Specifications
Specifications
Translucent
Autoclavable
1 7/8 in.
1/4 in.
1/4 in.
Autoclave or Ethylene Oxide
1/2 in.
Barb x NPT(M) Connection
PVDF
NPT Male Adapter to Hose Barb
1/4 in.
1/2 in.
1/2 in.