Learn More
VACUUBRAND™ PVDF/PTFE VV 15C In-Line Isolation Valve
Click to view available options
Size (Metric):
16 mm dia.
25 mm dia.
Unit Size:
Each
Specifications
Specifications
| Material | PVDF, PTFE |
| Quantity | 1 Each |
| Size (Metric) | 25 mm dia. |
| Type | In-Line Isolation Valve VV 15C |