missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PVRIG Antibody (2334B), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB93652L
This item is not returnable.
View return policy
Description
PVRIG Monoclonal antibody specifically detects PVRIG in Human samples. It is validated for Flow Cytometry
Specifications
| PVRIG | |
| Monoclonal | |
| DyLight 550 | |
| C7orf15MGC138295, MGC104322, MGC138297, MGC2463, poliovirus receptor related immunoglobulin domain containing, Poliovirus receptor-related immunoglobulin domain-containing protein, transmembrane protein PVRIG | |
| Human embryonic kidney cell line HEK293-derived recombinant human PVRIG, Thr41-Leu172, Accession # Q6DK17 | |
| 0.1 mL | |
| Cancer, Immunology | |
| 79037 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry | |
| 2334B | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction