missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PVRL1 Monoclonal Antibody (OTI6E11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503498S
This item is not returnable.
View return policy
Description
PVRL1 Monoclonal antibody specifically detects PVRL1 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| PVRL1 | |
| Monoclonal | |
| 0.73 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P23919 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1841 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI6E11 | |
| Unconjugated | |
| DTYMK | |
| AI835281; AW549174; CD111; CLPED1; ectodermal dysplasia 4 (Margarita Island type); ED4; herpes simplex virus type 1 sensitivity; herpes virus entry mediator C; Herpesvirus entry mediator C; herpesvirus Ig-like receptor; HIgR; HV1S; Hvec; nectin 1; nectin cell adhesion molecule 1; nectin); Nectin1; nectin-1; nectin-1 alpha; nectin-1 delta; OFC7; poliovirus receptor-like 1; poliovirus receptor-related 1; poliovirus receptor-related 1 (herpesvirus entry mediator C; poliovirus receptor-related 1 (herpesvirus entry mediator C); poliovirus receptor-related 1 (herpesvirus entry mediator C; nectin); poliovirus receptor-related protein 1; PRR; PRR1; PVRL1; PVRR; PVRR1; SK-12 | |
| Full length human recombinant protein of human DTYMK produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction