missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PVRL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102558
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PVRL1.
This gene encodes an adhesion protein that plays a role in the organization of adherens junctions and tight junctions in epithelial and endothelial cells. The protein is a calcium(2+)-independent cell-cell adhesion molecule that belongs to the immunoglobulin superfamily and has 3 extracellular immunoglobulin-like loops, a single transmembrane domain, and a cytoplasmic region. This protein acts as a receptor for glycoprotein D of herpes simplex viruses 1 and 2, and pseudorabies virus and mediates viral entry into epithelial and neuronal cells. Mutations in this gene cause cleft lip and palate/ectodermal dysplasia 1 syndrome as well as non-syndromic cleft lip with or without cleft palate. Alternative splicing results in multiple transcript variants encoding proteins with distinct C-termini.
Specifications
| PVRL1 | |
| Polyclonal | |
| Unconjugated | |
| Nectin1 | |
| AI835281; AW549174; CD111; CLPED1; ectodermal dysplasia 4 (Margarita Island type); ED4; herpes simplex virus type 1 sensitivity; herpes virus entry mediator C; Herpesvirus entry mediator C; herpesvirus Ig-like receptor; HIgR; HV1S; Hvec; nectin 1; nectin cell adhesion molecule 1; nectin); Nectin1; nectin-1; nectin-1 alpha; nectin-1 delta; OFC7; poliovirus receptor-like 1; poliovirus receptor-related 1; poliovirus receptor-related 1 (herpesvirus entry mediator C; poliovirus receptor-related 1 (herpesvirus entry mediator C); poliovirus receptor-related 1 (herpesvirus entry mediator C; nectin); poliovirus receptor-related protein 1; PRR; PRR1; PVRL1; PVRR; PVRR1; SK-12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 192183, 5818, 58235 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15223, Q9JKF6 | |
| Nectin1 | |
| A synthesized peptide derived from human NECTIN1(Accession Q15223), corresponding to amino acid residues N398-G448. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction