missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PVRL1 Rabbit anti-Human, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human PVRL1.
Brand: Abnova PAB29417.100uL
This item is not returnable.
View return policy
Description
Sequence: DSMYGFIGTDVVLHCSFANPLPSVKITQVTWQKSTNGSKQNVAIYNPSMGVSVLAPYRERVEFLRPSFTDGTIRLSRLELEDEGVYICEFATFPTGNRESQLNLTVMAKPTNWIEGTQAVLRAKKGQDDSpecifications
| PVRL1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant human PVRL1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| PVRL1 | |
| Recombinant protein corresponding to human PVRL1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| CD111/CLPED1/ED4/HIgR/HVEC/MGC142031/MGC16207/OFC7/PRR/PRR1/PVRR/PVRR1/SK-12/nectin-1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5818 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction