Learn More
PWP1 Monoclonal Antibody (3A11)
PWP1 Monoclonal Antibody (3A11)
Brand: Invitrogen MA521268
This item is not returnable.
View return policy
Description
PWP1 Monoclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
| PWP1 | |
| Monoclonal | |
| 0.42 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q13610 | |
| PWP1 | |
| PWP1 (AAH01652, 1 a.a. ~ 501 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 3A11 | |
| Unconjugated | |
| PWP1 | |
| 2310058A11Rik; 2610205J09Rik; AI098170; AL022879; AV083409; AW550690; C20orf152; endonuclein; IEF-SSP-9502; Keratinocyte protein IEF SSP 9502; nuclear phosphoprotein similar to S. cerevisiae PWP1; periodic tryptophan protein 1 homolog; Pwp1; PWP1 homolog (S. cerevisiae); PWP1 homolog, endonuclein; RGD1310705 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11137 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |