missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PWP1 Recombinant Rabbit Monoclonal Antibody (23GB2580), Invitrogen™
GREENER_CHOICE

Product Code. p-200087634
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30089248 100 μL 100µL
30087930 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30089248 Supplier Invitrogen™ Supplier No. MA553556

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

The protein encoded by this gene contains several WD-40 repeats and is found mostly in the nucleus. The expression and localization of this protein are cell cycle dependent. Expression of this gene is upregulated in pancreatic adenocarcinoma. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PWP1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2580
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PWP1
Gene Accession No. Q13610, Q99LL5
Gene Alias 2310058A11Rik; 2610205J09Rik; AI098170; AL022879; AV083409; AW550690; C20orf152; endonuclein; IEF-SSP-9502; Keratinocyte protein IEF SSP 9502; nuclear phosphoprotein similar to S. cerevisiae PWP1; periodic tryptophan protein 1 homolog; Pwp1; PWP1 homolog (S. cerevisiae); PWP1 homolog, endonuclein; RGD1310705
Gene Symbols PWP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PWP1 (460-501AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103136, 11137
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.