Learn More
Invitrogen™ PWWP2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140013
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VRVEQVVNGA LVVTVSCGER SFAGILLDCT KKSGLFGLPP LAPLPQVDES PVNDSHGRAP EEGDAEVMQL Highest antigen sequence indentity to the following orthologs: Rat - 77%, Mouse - 79%.
PWWP2B is a protein coding gene.
Specifications
| PWWP2B | |
| Polyclonal | |
| Unconjugated | |
| PWWP2B | |
| bA432J24.1; pp8607; PWWP domain containing 2; PWWP domain containing 2B; PWWP domain-containing protein 2B; PWWP2; PWWP2B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 170394 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NUJ5 | |
| PWWP2B | |
| Recombinant protein corresponding to Human PWWP2B. Recombinant protein control fragment (Product #RP-109668). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.