missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PXDN, Mouse anti-Human, Clone: 2C11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00007837-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: STSAFSTRSDASGTNDFREFVLEMQKTITDLRTQIKKLESRLSTTECVDAGGESHANNTKWKKDACTICECKDGQVTCFVEACPPATCAVPVNIPGACCPVCLQKRAEEKSpecifications
| PXDN | |
| Monoclonal | |
| Unconjugated | |
| peroxidasin homolog (Drosophila) | |
| D2S448/D2S448E/KIAA0230/MG50/PRG2/PXN/VPO | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2C11 | |
| In 1x PBS, pH 7.4 | |
| XM_056455 | |
| PXDN | |
| PXDN (XP_056455, 1452 a.a. ∼ 1561 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 7837 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction