missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PXMP4 Antibody (7S41), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-21987H
This item is not returnable.
View return policy
Description
PXMP4 Monoclonal specifically detects PXMP4 in Mouse samples. It is validated for ELISA.
Specifications
| PXMP4 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| PXMP4 | |
| Mouse recombinant PXMP4 | |
| 0.1 mL | |
| 11264 | |
| Store at 4C in the dark. | |
| IgG2 |
| ELISA | |
| 7S41 | |
| ELISA | |
| PMP24, PXMP4 peroxisomal membrane protein 4, 24kDa | |
| Rat | |
| Protein G purified | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction