Learn More
PXN Monoclonal Antibody (OTI2B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA811258
Encompass_Preferred
 View more versions of this product
Product Code. 17535114
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.
Specifications
Specifications
PXN
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P49023
PXN
Full length human recombit protein of human PXN produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI2B8
Unconjugated
PXN
paxillin
Mouse
Affinity Chromatography
RUO
5829
-20° C, Avoid Freeze/Thaw Cycles