missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PXR/NR1I2 Antibody (V11P4G11*E7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00 - € 407.00
Specifications
| Antigen | PXR/NR1I2 |
|---|---|
| Clone | V11P4G11*E7 |
| Concentration | 1.1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18655958
|
Novus Biologicals
NBP2-50519-0.025ml |
0.025 mL |
€ 190.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18614139
|
Novus Biologicals
NBP2-50519 |
0.1 mL |
€ 407.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PXR/NR1I2 Monoclonal antibody specifically detects PXR/NR1I2 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| PXR/NR1I2 | |
| 1.1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Stem Cell Markers, Transcription Factors and Regulators | |
| PBS | |
| 8856 | |
| IgG1 κ | |
| Protein A purified |
| V11P4G11*E7 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Zebrafish | |
| nuclear receptor subfamily 1 group I member 2, nuclear receptor subfamily 1, group I, member 2, ONR1, Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, PAR, PAR1, PAR2, Pregnane X receptor, PRR, PXRPARq, SAR, Steroid and xenobiotic receptor, SXRBXR | |
| Ovalbumin-conjugated synthetic peptide EMRSMNEEYT (C-terminal sequence) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title