missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYCARD Monoclonal Antibody (OTI1A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800079S
This item is not returnable.
View return policy
Description
PYCARD Monoclonal antibody specifically detects PYCARD in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| PYCARD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30740 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1992 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1A2 | |
| Unconjugated | |
| SERPINB1 | |
| 9130417A21Rik; Apoptosis-associated speck-like protein containing a CARD; Asc; CARD5; caspase recruitment domain-containing protein 5; hASC; mASC; MGC10332; Pycard; PYD and CARD domain containing; PYD and CARD domain-containing protein; Target of methylation-induced silencing 1; TMS; TMS1; TMS-1; TNS1 | |
| Human recombinant protein fragment corresponding to amino acids 77-306 of human SERPINB1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction