Learn More
PYCR2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-17146-20UL
Description
The PYCR2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PYCR2. This antibody recognizes human, mouse, rat antigen. The PYCR2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| PYCR2 | |
| 1000 μg/mL | |
| FLJ54750, P5C reductase 2, P5CR 2, P5CR2, PYCR2 | |
| 20 μL | |
| 29920 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |