missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PYCRL Monoclonal Antibody (OTI1B12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525335
This item is not returnable.
View return policy
Description
PYCRL Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
PYCRL encodes a protein that belongs to the pyrroline-5-carboxylate reductase family of enzymes. Members of this family catalyze the final step in proline biosynthesis, converting pyrroline-5-carboxylate to proline. Glutamate and ornithine are precursors in the synthesis of proline. The protein encoded by this gene is a cytoplasmic enzyme involved in the biosynthesis of proline from ornithine.
Specifications
| PYCRL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q53H96, Q5PQJ6 | |
| PYCR3 | |
| Full length human recombinant protein of PYCRL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1B12 | |
| Unconjugated | |
| PYCR3 | |
| 1110058B13Rik; 2700073G24Rik; P5C reductase 3; P5CR 3; PYCR3; Pycrl; pyrroline-5-carboxylate reductase 3; pyrroline-5-carboxylate reductase-like; pyrroline-5-carboxylate reductase-like protein | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 300035, 65263 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction