missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYCRL Monoclonal Antibody (OTI3A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502032
This item is not returnable.
View return policy
Description
PYCRL encodes a protein that belongs to the pyrroline-5-carboxylate reductase family of enzymes. Members of this family catalyze the final step in proline biosynthesis, converting pyrroline-5-carboxylate to proline. Glutamate and ornithine are precursors in the synthesis of proline. The protein encoded by this gene is a cytoplasmic enzyme involved in the biosynthesis of proline from ornithine.Specifications
| PYCRL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q53H96 | |
| PYCR3 | |
| Full length human recombit protein of human PYCRL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI3A8 | |
| Unconjugated | |
| PYCR3 | |
| PYCRL | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 65263 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction