missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYDC1 Monoclonal Antibody (OTI5D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803566
This item is not returnable.
View return policy
Description
ASC2 associates with PYCARD/ASC and modulates its ability to collaborate with MEFV/pyrin and NLRP3/cryopyrin in NF-kappa-B and pro-caspase-1 activation. It suppresses kinase activity of NF-kappa-B inhibitor kinase (IKK) complex, expression of NF-kappa-B inducible genes and inhibits NF-kappa-B activation by cytokines and LPS.Specifications
| PYDC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WXC3 | |
| PYDC1 | |
| Full length human recombit protein of human PYDC1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5D12 | |
| Unconjugated | |
| PYDC1 | |
| ASC2, POP1, PYC1, cPOP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 260434 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction