missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYDC1 Mouse anti-Human, Clone: OTI9C10, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803466S
This item is not returnable.
View return policy
Description
PYDC1 Monoclonal antibody specifically detects PYDC1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PYDC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WXC3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 260434 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9C10 | |
| Unconjugated | |
| PYDC1 | |
| ASC2; ASCI; cellular POP1; cPOP1; PAAD-only protein 1; POP1; PYC1; PYD (pyrin domain) containing 1; PYDC1; pyrin domain containing 1; pyrin domain-containing protein 1; Pyrin-only protein 1 | |
| Full length human recombinant protein of human PYDC1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction