missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PYGB Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121953
This item is not returnable.
View return policy
Description
PYGB Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is a glycogen phosphorylase found predominantly in the brain. The encoded protein forms homodimers which can associate into homotetramers, the enzymatically active form of glycogen phosphorylase. The activity of this enzyme is positively regulated by AMP and negatively regulated by ATP, ADP, and glucose-6-phosphate. This enzyme catalyzes the rate-determining step in glycogen degradation.
Specifications
| PYGB | |
| Polyclonal | |
| Unconjugated | |
| PYGB | |
| brain glycogen phosphorylase; glycogen phosphorylase B; glycogen phosphorylase, brain form; GLYPHOA; GPBB; MGC9213; phosphorylase, glycogen; brain; PYGB | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 110078, 5834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P11216, Q8CI94 | |
| PYGB | |
| Recombinant protein Glycogen Phosphorylase, Brain. The antigen corresponds to amino acid range 477-731 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction