missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYGM Monoclonal Antibody (OTI3F9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811315
This item is not returnable.
View return policy
Description
PYGM encodes a muscle enzyme involved in glycogenolysis. Highly similar enzymes encoded by different genes are found in liver and brain. Mutations in this gene are associated with McArdle disease (myophosphorylase deficiency), a glycogen storage disease of muscle. Alternative splicing results in multiple transcript variants.Specifications
| PYGM | |
| Monoclonal | |
| Unconjugated | |
| PYGM | |
| glycogen phosphorylase, muscle associated | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5837 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3F9 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11217 | |
| PYGM | |
| Human recombit protein fragment corresponding to amino acids 698-842 of human PYGM produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction