Learn More
Pyrazole-1-propionitrile (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | Pyrazole-1-propionitrile |
| CAS | 88393-88-8 |
| Molecular Formula | C6 H7 N3 |
| Quantity | 100 mg |
| Synonym | 3-(1H-Pyrazol-1-yl)propanenitrile, 3-Pyrazol-1-ylpropanenitrile |
| InChI Formula | InChI=1S/C6H7N3/c7-3-1-5-9-6-2-4-8-9/h2,4,6H,1,5H2 |
| SMILES | N#CCCn1cccn1 |
| IUPAC Name | 3-pyrazol-1-ylpropanenitrile |
| Molecular Weight (g/mol) | 121.14 |
| Formula Weight | 121.064 |