missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid, 95%, Thermo Scientific™

Product Code. 15179303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15179303 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15179303 Supplier Thermo Scientific Acros Supplier No. 449920010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 25940-35-6
Melting Point 275.0°C to 280.0°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H5N3O2
MDL Number MFCD01975194
Quantity 1 g
Synonym pyrazolo 1,5-a pyrimidine-3-carboxylic acid, pyrazolo 1,5-a pyrimidine-3-carboxylicacid, 3-carboxypyrazolo 1,5-a pyrimidine, 8-hydropyrazolo 1,5-a pyrimidine-3-carboxylic acid, pubchem20936, acmc-1cbw6, 3-pyrazolo 1,5-a pyrimidinecarboxylic acid
InChI Key HNYVPKNVKSTVJO-UHFFFAOYSA-N
SMILES OC(=O)C1=C2N=CC=CN2N=C1
IUPAC Name pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
Molecular Weight (g/mol) 163.14
PubChem CID 1910189
Formula Weight 163.13
Percent Purity 95%
Chemical Name or Material Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.