Learn More
Pyrethroid hydrolase Ces2e Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-06082PE
Product Code. 18699218
  • € 407.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Pyrethroid hydrolase Ces2e Polyclonal antibody specifically detects Pyrethroid hydrolase Ces2e in Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Pyrethroid hydrolase Ces2e
Polyclonal
Western Blot, ELISA
9030624L02Rik, ACH M1;CE-2;CES 2;CES2;CES2A1;ces2A3;Ces2c;Ces2e;ICE;PCE-2, carboxylesterase 5, carboxylic ester hydrolase, Ces5
Produced in rabbits immunized with purified, recombinant Mouse Pyrethroid hydrolase Ces2e (Accession#: NP_766347.1; Met1-His556)
0.1 mL
Immunology
234673
Store at 4C in the dark.
IgG
Western Blot, ELISA
PE
PBS
Rabbit
Antigen and protein A Affinity-purified